Anti-BG-9588 Recombinant Antibody [clone: 2196]
Catalog # PRSI10-064
Supplier: ProSci Inc.
Specifications
- Antibody type:Primary
- Clonality:Recombinant
- Clone:2196
- Conjugation:Unconjugated
- ELISA:Yes
- Reactivity:Virus
- Form:Liquid
- Antigen synonyms:Receptor Binding Domain Antibody|SARS-CoV-2 Spike RBD Antibody
- Storage buffer:This monoclonal antibody is aseptically packaged and formulated in 0,01 M phosphate buffered saline (PBS) pH 7,2 - 7,4, 150 mM NaCl with no carrier protein, potassium, calcium or preservatives added
- Concentration:>1,0 mg/ml
- Shipping temperature:20
- Immunogen:Sequenced from human survivors of COVID-19 (SARS-CoV-2). The spike RBD is expressed on the surface of SARS-CoV-2.
- Purification:>95% by SDS page
- Pack type:Vial
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen:
Clonality: Recombinant
Clone: 2196
Conjugation: Unconjugated
Epitope:
Host:
Isotype: Human IgG1
Reactivity: Virus