Order Entry
Austria
ContactUsLinkComponent
Anti-Adgre1 Recombinant Antibody [Clone: Cl:A3-1 (recombinant version)]
Anti-Adgre1 Recombinant Antibody [Clone: Cl:A3-1 (recombinant version)]
Catalog # PRSI24-110
Supplier:  ProSci Inc.
Anti-Adgre1 Recombinant Antibody [Clone: Cl:A3-1 (recombinant version)]
Catalog # PRSI24-110
Supplier:  ProSci Inc.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    adhesion G protein-coupled receptor E1
  • Antigen symbol:
    Adgre1
  • Clonality:
    Recombinant
  • Clone:
    Cl:A3-1 (recombinant version)
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2b kappa
  • Cross adsorption:
    No
  • Form:
    Liquid
  • Gene ID:
    13733
  • Antigen synonyms:
    f-480|EMR1 receptor|mouse macrophage marker|EGF-like module receptor 1|Em Gpf4|f 480|f480|f4-80|Ly-71|Cell surface glycoprotein F4/80|f4 80|TM7|f-4/80|EGF-like module
  • Modification:
    Unmodified
  • Storage buffer:
    PBS with 0,02% Proclin 300
  • Storage temperature:
    Store at 4 °C for up to 3 months. For longer storage, aliquot and store at −20 °C.
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Thioglycollate stimulated peritoneal macrophages of mouse origin
  • Pack type:
    Vial
  • Pk:
    0,2 mg

Specifications

About this item

Type: Primary
Antigen: adhesion G protein-coupled receptor E1
Clonality: Recombinant
Clone: Cl:A3-1 (recombinant version)
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG2b kappa
Reactivity: Mouse