Order Entry
Austria
Orders LinkContactUsLinkComponent
Formalin 10% histological fixative, neutral buffered

Formalin 10% histological fixative, neutral buffered

Irritant
Health
Formalin 10% histological fixative, neutral buffered
Certificates
Formalin 10% histological fixative, neutral buffered

2 Options Available Below

Product Details & Documents

10% Neutral buffered formalin (NBF) is the most commonly used fixative for light microscopy. 10% NBF is well suited for large throughput laboratories and requires a relatively short period of fixation, but can also be used for the long-term storage of tissue. Formalin fixation is thought to form between the aldehydes and the proteins, creating a gel, thus retaining cellular constituents in their in vivo relationship.

  • Formalin, 10%, Neutral Buffered, Wintergreen Scented has been used as a fixative in transformation and tumorigenicity assay[3] and in immunohistostaining
  • Intended for use as a general purpose histological fixative
  • Wintergreen oil has been added to the standard formulation to provide a wintergreen scent
Synonyms:  Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  •  :M-Clarity™ quality level = MQ200
  • Storage Temperature:Room Temperature

Documents

Specifications for Product Family

Specification Test Results

  • Appearance (Colour):
    Colorless
  • Appearance (Form):
    Liquid
  • Appearance (Turbidity):
    Clear
  • pH:
    6.9 - 7.1
  • Assay
    % Formaldehyde (w/v):
    3.5 - 4.2 %
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 4 L
    Log in for availability
    Log in for availability
    1 * 4 L
    1 * 4 L - 66,90 €
    66,90 €
    1 * 4 L
       66,90 €

  • 19 L
    Log in for availability
    Log in for availability
    1 * 19 L
    1 * 19 L - 193,00 €
    193,00 €
    1 * 19 L
       193,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.