Order Entry
Austria
ContactUsLinkComponent
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) fluorescent labeling reagent
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) fluorescent labeling reagent
Certificates
undefined
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) fluorescent labeling reagent

Specification Test Results

  • Appearance (Colour):
    Yellow to Orange
  • Appearance (Form):
    Powder
  • Assay
    FTIC Content:
    8.0 - 12.0 %
  • Suitability
    Suitable for use in Protein Labeling:
    Suitable

Specification Test Results

Frequently Bought Together