Order Entry
Austria
ContactUsLinkComponent
 
Pipette tips, uTIP™, X-Resin™
 :  BIOTIX
    
 
undefined
Pipette tips, uTIP™, X-Resin™
 :  BIOTIX
    
 

 

  • Catalog No:
  • 613-5863
  • 613-5861
  • BITX63300075
  • BITX63300073
  • 613-2814
  • 613-5865
  • BITX63300054
  • 613-5613
  • BITX63300060
  • 613-5615
  • 613-5616
  • 613-5619
  • BITX63300059
  • 613-2813
  • BITX63300071
  • 613-5866
  • 613-5862
  • BITX63300079
  • 613-5647
  • BITX63300080
  • 613-2815
  • BITX63300048
  • BITX63300061
  • 613-5624
  • 613-5612
  • 613-5622
  • BITX63300072
  • BITX63300050
  • BITX63300058
  • BITX63300051
  • 613-1377
  • 613-1379
  • 613-5623
  • Environmentally Preferable:
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Environmental certification:
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C1544854/30/2025; Bulk tips reduce plastic waste by 95%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to individually racked products; Bulk tips reduce plastic waste by 95%
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Bulk tips reduce plastic waste by 95%

 

Empty racks

 

Frequently Bought Together