Order Entry
Austria
Orders LinkContactUsLinkComponent
 

Eppendorf® DNA LoBind Plates

   Sustainable Options Available
Supplier:   EPPENDORF
Eppendorf® DNA LoBind Plates
Supplier:  EPPENDORF
   Sustainable Options Available
Certificates
Eppendorf® DNA LoBind Plates

10 Options Available Below

Product Details & Documents

DNA LoBind Plates maximise sample recovery of nucleic acids by significantly reducing sample-to-surface binding. These plates are the ideal solution for sample preparation and long-term storage.

  • Eppendorf LoBind material ensures optimised sample recovery for improved assay results
  • Free of surface coating (e.g. silicone) to minimise the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • High-contrast Unique OptiTrack® matrix: up to 30% faster sample identification and fewer pipetting errors
  • RecoverMax® well design: Optimised well geometry for minimal remaining/dead volume and excellent mixing properties
  • Raised well rims and a smooth surface ensure reliable sealing

Maximise recovery of nucleic acids can be achieved with DNA LoBind® plates. A combination of special manufacturing technologies and selected polypropylene batches ensure nearly 100 % recovery of DNA/RNA molecules-without surface coating to eliminate the risk of sample contamination. This makes Eppendorf LoBind Plates ideal for applications where sample recovery is key - such as the preparation of long-term storage nucleic acids in forensic, microarrays and NGS applications.

Purity grade: PCR Clean
Material: Polypropylene (PP)
Autoclavable: 121 °C, 20 min
Centrifugation stability: 6000×g
Well colour: Clear

Specifications for Product Family

Specifications

  • Catalog No:
  • 737-0214
  • 737-2530
  • 737-2534
  • 737-0206
  • 737-2535
  • 737-2539
  • 737-0207
  • 737-0162
  • 737-2559
  • 737-0212
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001

Specifications

Product Family Options

96-well Plates

Product Information

  • Colour
    Packed
    Well volume
    Availability
    Price
  • 737-0214
    Microplate
    Microplate
    White
    80 plates (5 bags × 16 plates)
    350 µl
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 273,00 €
    273,00 €
    1 * 80 ST
       273,00 €

    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0214
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • Deepwell plate
    White
    40 plates (5 bags × 8 plates)
    500 µl
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 435,00 €
    435,00 €
    1 * 40 ST
       435,00 €

    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-2530
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-2534
    Deepwell plate
    Deepwell plate
    Blue
    40 plates (5 bags × 8 plates)
    500 µl
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 435,00 €
    435,00 €
    1 * 40 ST
       435,00 €

    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-2534
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-0206
    Deepwell plate
    Deepwell plate
    White
    120 plates (10 bags × 12 plates)
    500 µl
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 1 174,00 €
    1 174,00 €
    1 * 120 ST
       1 174,00 €

    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0206
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-2535
    Deepwell plate
    Deepwell plate
    White
    20 plates (5 bags × 4 plates)
    1000 µl
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 198,00 €
    198,00 €
    1 * 20 ST
       198,00 €

    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-2535
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-2539
    Deepwell plate
    Deepwell plate
    Blue
    20 plates (5 bags × 4 plates)
    1000 µl
    Log in for availability
    Log in for availability
    1 * 20 ST
    1 * 20 ST - 198,00 €
    198,00 €
    1 * 20 ST
       198,00 €

    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-2539
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-0207
    Deepwell plate
    Deepwell plate
    White
    80 plates (10 bags × 8 plates)
    1000 µl
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 711,00 €
    711,00 €
    1 * 80 ST
       711,00 €

    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0207
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact

384-well Plates

Product Information

  • Colour
    Packed
    Well volume
    Availability
    Price
  • 737-0162
    Microplate
    Microplate
    White
    80 plates (5 bags × 16 plates)
    140 µl
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 746,00 €
    746,00 €
    1 * 80 ST
       746,00 €

    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0162
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-2559
    Deepwell plate
    Deepwell plate
    White
    40 plates (5 bags × 8 plates)
    200 µl
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 490,00 €
    490,00 €
    1 * 40 ST
       490,00 €

    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-2559
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-0212
    Deepwell plate
    Deepwell plate
    White
    120 plates (10 bags × 12 plates)
    200 µl
    Log in for availability
    Log in for availability
    1 * 120 ST
    1 * 120 ST - 1 318,00 €
    1 318,00 €
    1 * 120 ST
       1 318,00 €

    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0212
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact