Order Entry
Austria
ContactUsLinkComponent
Anti-SP Rabbit Polyclonal Antibody
Anti-SP Rabbit Polyclonal Antibody
Catalog # ENZOBMLSA12700100
Supplier:  ENZO LIFE SCIENCES
Anti-SP Rabbit Polyclonal Antibody
Catalog # ENZOBMLSA12700100
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antigen name:
    Substance P
  • Antigen symbol:
    SP
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Seq:
    02
  • Format:
    Liquid, antiserum containing 0,1% sodium azide
  • UniProtKB:
    P20366
  • Storage temperature:
    −20 °C
  • Shipping temperature:
    20
  • Immunogen:
    Synthetic substance P (H-RPKPQQFFGLM-NH2)
  • Tested applications:
    IHC
  • Pk:
    100 µl

Specifications

About this item

Type:
Antigen: SP
Clonality: Polyclonal 
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Guinea pig, Human, Porcine, Rat