Orders LinkContactUsLinkComponent
Eppendorf® Microplates
Eppendorf® Microplates
   Sustainable Options Available
Supplier:   EPPENDORF
Eppendorf® Microplates
Supplier:  EPPENDORF
   Sustainable Options Available
Certificates
Eppendorf® Microplates
13 Options Available Below
Product Details & Documents
Eppendorf microplates provide exceptional stability during centrifugation with g-Safe® technology and efficient sample identification with the OptiTrack® Matrix. The design features, including RecoverMax® well design and raised well rims, ensure reliable sealing and facilitate maximum sample recovery.
  • g-Safe®: exceptional centrifugation stability up to 6000×g
  • OptiTrack® Matrix: up to 30% faster sample identification and fewer pipetting errors via high-contrast alphanumeric labelling
  • RecoverMax® well design: Optimised well geometry for minimal remaining volume and excellent mixing properties
  • Raised well rims and smooth surface for reliable closing, even with heat sealing
  • Unmatched, transparent polypropylene (PP) for improved sample visibility
  • Polypropylene ensures high resistance to chemicals, mechanical stress and temperature extremes
  • Manufactured without slip agents, plastocizers or biocides (materials which could have a negative effect on bioassays)
  • Ensured functionality from −86 to +100 °C
Pipette, control and recover your samples with ease using Eppendorf Microplates! Designed with the easy-to-read alphanumeric OptiTrack® matrix label to identify individual wells, speed up the filling process and significantly reduce the risk of pipetting errors. Eppendorf Microplates are made with high-transparency polypropylene ensuring quick and easy location of samples and are also available in Eppendorf LoBind® for when you need maximum sample recovery.
Safe code option provides 3-level coding for safe sample and vessel identification. ECC 200 error correction function enables safe code reading with up to 30% damaged codes.
Well colour: Clear
Material: polypropylene (PP)
Autoclavable: 121 °C, 20 min
Centrifugation stability: 6000×g
Plate colour: white
Specifications for Product Family
Specifications
  • Catalog No:
  • 737-0166
  • 737-0167
  • 737-0168
  • 737-0254
  • 737-0173
  • 737-0174
  • 737-0175
  • 737-0224
  • 737-0255
  • 737-0225
  • 737-0226
  • 737-0229
  • 737-0230
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
  • Renewable Energy Certificate, ISO 14001
Specifications
Product Family Options
96-well
Product Information
  • No. of wells
    Sterile
    Packed
    Availability
    Price
  • 737-0166
    Eppendorf® Microplate, flat bottom, PCR clean (Non sterile)
    Eppendorf® Microplate, flat bottom, PCR clean (Non sterile)
    -
    80 plates (5 bags × 16 plates)
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 254,00 €
    254,00 €
    1 * 80 ST
       254,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0166
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-0167
    Eppendorf® Microplate, U-bottom, PCR clean (Non sterile)
    Eppendorf® Microplate, U-bottom, PCR clean (Non sterile)
    96
    -
    80 plates (5 bags × 16 plates)
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 254,00 €
    254,00 €
    1 * 80 ST
       254,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0167
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-0168
    Eppendorf® Microplate, V-bottom, PCR clean (Non sterile)
    Eppendorf® Microplate, V-bottom, PCR clean (Non sterile)
    96
    -
    80 plates (5 bags × 16 plates)
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 254,00 €
    254,00 €
    1 * 80 ST
       254,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0168
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-0254
    Eppendorf® Microplate, 3-level SafeCode (2D datamatrix, linear barcode, alphanumeric code), V-bottom, PCR clean (Non sterile)
    Eppendorf® Microplate, 3-level SafeCode (2D datamatrix, linear barcode, alphanumeric code), V-bottom, PCR clean (Non sterile)
    -
    80 plates (5 bags × 16 plates)
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 384,00 €
    384,00 €
    1 * 80 ST
       384,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0254
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-0173
    Eppendorf® Microplate, flat bottom
    Eppendorf® Microplate, flat bottom
    96
    +
    80 plates (5 bags × 16 plates)
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 291,00 €
    291,00 €
    1 * 80 ST
       291,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0173
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-0174
    Eppendorf® Microplate, U-bottom
    Eppendorf® Microplate, U-bottom
    96
    +
    80 plates (5 bags × 16 plates)
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 291,00 €
    291,00 €
    1 * 80 ST
       291,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0174
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-0175
    Eppendorf® Microplate, V-bottom
    Eppendorf® Microplate, V-bottom
    96
    +
    80 plates (5 bags × 16 plates)
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 291,00 €
    291,00 €
    1 * 80 ST
       291,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0175
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-0224
    Eppendorf® Microplate, V-bottom
    Eppendorf® Microplate, V-bottom
    +
    240 plates (10 bags × 24 plates)
    Log in for availability
    Log in for availability
    1 * 240 ST
    1 * 240 ST - 783,00 €
    783,00 €
    1 * 240 ST
       783,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    737-0224
    Environmentally Preferable
    Environmental certification
384-well
Product Information
  • No. of wells
    Sterile
    Packed
    Availability
    Price
  • 737-0255
    Eppendorf® Microplate, 3-level SafeCode (2D datamatrix, linear barcode, alphanumeric code), V-bottom, PCR clean (Non sterile)
    Eppendorf® Microplate, 3-level SafeCode (2D datamatrix, linear barcode, alphanumeric code), V-bottom, PCR clean (Non sterile)
    -
    80 plates (5 bags × 16 plates)
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 760,00 €
    760,00 €
    1 * 80 ST
       760,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0255
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-0225
    Eppendorf® Microplate, flat bottom, PCR clean (Non sterile)
    Eppendorf® Microplate, flat bottom, PCR clean (Non sterile)
    384
    -
    80 plates (5 bags × 16 plates)
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 645,00 €
    645,00 €
    1 * 80 ST
       645,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0225
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-0226
    Eppendorf® Microplate, V-bottom, PCR clean (Non sterile)
    Eppendorf® Microplate, V-bottom, PCR clean (Non sterile)
    384
    -
    80 plates (5 bags × 16 plates)
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 645,00 €
    645,00 €
    1 * 80 ST
       645,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0226
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-0229
    Eppendorf® Microplate, flat bottom
    Eppendorf® Microplate, flat bottom
    384
    +
    80 plates (5 bags × 16 plates)
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 746,00 €
    746,00 €
    1 * 80 ST
       746,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0229
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact
  • 737-0230
    Eppendorf® Microplate, V-bottom
    Eppendorf® Microplate, V-bottom
    384
    +
    80 plates (5 bags × 16 plates)
    Log in for availability
    Log in for availability
    1 * 80 ST
    1 * 80 ST - 746,00 €
    746,00 €
    1 * 80 ST
       746,00 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    737-0230
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    Renewable Energy Certificate, ISO 14001
      Low Manufacturing Impact