ContactUsLinkComponent
Wash Solution Concentrate, 20X
Wash Solution Concentrate, 20X
Catalog # KPLI50-63-04
CAS Number:  
Wash Solution Concentrate, 20X
Catalog # KPLI50-63-04
CAS Number:  
Specifications
  • Pack type:
    Bottle
  • Pk:
    1 L
  • Description:
    20X wash solution concentrate
  • Seq:
    02
  • Environmentally Preferable:
  • Pk Info:
    1×1 L
Specifications
About this item
Convenient 20X concentrate containing imidazole-buffered saline and Tween® 20.
  • Suitable for ELISA, blotting, and immunohistochemistry applications
This is used a general immunoassay wash buffer.