Sodium acetate ≥99.0% ACS
SIAL241245-100G
: Merck
:
- Pack type:Plastic bottle, wide mouth
- Pk:100 g
- Seq:3401
- Assay:99.0%
:
:82.03 g/mol
:12,08711
:881,4 °C (1013 hPa)
:324 °C
:1,528 g/cm³ (20 °C)
:>250 °C (closed cup)
:Ambient
:204-823-8
:01-2119485123-42
:127-09-3
:H₃CCOONa
:MFCD00012459