Orders LinkContactUsLinkComponent
Gel tools
Gel tools
Gel tools
Certificates
Gel tools
6 Options Available Below
Product Details & Documents
Gel cutter: The gel cutter's edge allows for easy cutting and removal of gel material.
Gel ruler: The gel ruler has centimetre markings that fluoresce under 365 and 302 nm UV wavelengths, providing reference marks for DNA analysis.
Gel scooper: Made of strong acrylic with a bevelled edge, the gel scooper is designed for easy transfer of gels from electrophoresis equipment to viewing equipment.
Gel trays: Gel trays (in two sizes), made of UV transmitting Plexiglass, can be used for moving gels to the transilluminator. The tray protects the transilluminator filter surface from scratches. Side panels on the gel tray, angled at 45°, extend upward from the tray surface for easy handling.
Fluorescent standard step tablet: The step tablet has a calibrated 21-step gradation with density from totally clear (step 1) to totally opaque (step 21). The gradation on the step tablet allows accurate, consistent comparison of the bands and spots on acquired images for variations in lighting and optical conditions and for maximising fluorescence calibration of images. The guide lets researchers evaluate the image against the values of grey, maximising fluorescence calibration and providing a standard reference for images.
Product Family Options
Product Information
  • Availability
    Price
  • 700-1440
    Gel cutter, 200 mm long
    Gel cutter, 200 mm long
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,90 €
    23,90 €
    1 * 1 ST
       23,90 €
  • 700-1444
    Gel ruler, 250 mm long
    Gel ruler, 250 mm long
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,30 €
    11,30 €
    1 * 1 ST
       11,30 €
  • 730-1022
    Gel scooper, 127×305 mm
    Gel scooper, 127×305 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 94,20 €
    94,20 €
    1 * 1 ST
       94,20 €
  • 730-1021
    Gel tray, 420×270 mm
    Gel tray, 420×270 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 102,00 €
    102,00 €
    1 * 1 ST
       102,00 €
  • 730-1023
    Gel tray, 290×229 mm
    Gel tray, 290×229 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 74,70 €
    74,70 €
    1 * 1 ST
       74,70 €
  • 733-2011
    Fluorescent standard step tablet
    Fluorescent standard step tablet
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 443,00 €
    443,00 €
    1 * 1 ST
       443,00 €