Order Entry
Austria
ContactUsLinkComponent
Pig Trypsin (from Pancreas)
Pig Trypsin (from Pancreas)
Catalog # SIALT5266-500MG
Supplier:  Merck
CAS Number:  
Pig Trypsin (from Pancreas)
Catalog # SIALT5266-500MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • CAS No.:
    9002-07-7
  • Source:
    Pancreas
  • Species:
    Pig
  • Storage conditions:
    Refrigerator
  • MDL No.:
    MFCD00082094
  • Enzyme name:
    Trypsin

Specifications

About this item