Order Entry
Austria
ContactUsLinkComponent
VWR® Native Bovine Trypsin (from Pancreas)
VWR® Native Bovine Trypsin (from Pancreas)
Catalog # M150-1G
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Native Bovine Trypsin (from Pancreas)
Catalog # M150-1G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • CAS No.:
    9002-07-7
  • Source:
    Pancreas
  • Species:
    Bovine
  • Storage conditions:
    Refrigerator
  • Form:
    Crystals
  • Enzyme name:
    Trypsin
  • Enzyme activity:
    2.500 U/mg
  • Grade:
    Proteomics grade

Specifications

About this item

From bovine pancreas.