Order Entry
Austria
ContactUsLinkComponent
Mercury(II) nitrate 0.01 mol/l (0.02 N) in aqueous solution
Health
Irritant
Mercury(II) nitrate 0.01 mol/l (0.02 N) in aqueous solution
Catalog # 310274.1000
Supplier:  VWR Chemicals
CAS Number:  
undefined
Health
Irritant
Mercury(II) nitrate 0.01 mol/l (0.02 N) in aqueous solution
Catalog # 310274.1000
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :324.60 g/mol
EINECS:233-152-3
UN:3287
ADR:6.1,II
 :GHS08
 :GHS07
CAS Number:10045-94-0
Formula:HgN₂O₆
MDL Number:MFCD00011038