Order Entry
Austria
ContactUsLinkComponent
Mercury(II) nitrate 0.005 M Hg(NO3)2 (0.01N) ≥98% volumetric solution, for chloride determination, Fluka™
Health
Irritant
Mercury(II) nitrate 0.005 M Hg(NO3)2 (0.01N) ≥98% volumetric solution, for chloride determination, Fluka™
Catalog # 34289-1L
CAS Number:  
Health
Irritant
Mercury(II) nitrate 0.005 M Hg(NO3)2 (0.01N) ≥98% volumetric solution, for chloride determination, Fluka™
Catalog # 34289-1L
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Factor:
    0.995 - 1.005

Specification Test Results

About this item

 :
 :324.60 g/mol
Boiling Pt:100 °C (solvent)
Melting Pt:<0 °C
Density:1.0 g/cm³ (20 °C)
Flash Pt:Does not flash
Flash Pt: Does not flash
EINECS:233-152-3
ADR:6.1,II
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:10045-94-0
Formula:HgN₂O₆
MDL Number:MFCD00011038