Phenol red sodium salt 0.02% (w/v) in aqueous solution
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:34487-61-1
:
Formula:C₁₉H₁₃NaO₅S
MDL Number:MFCD00066901
:376.35 g/mol
Density:0.972
Storage Temperature:Ambient
EINECS:252-057-8
Related Information
Frequently Bought Together