Order Entry
Austria
ContactUsLinkComponent
Sodium chloride 26% in aqueous solution
Sodium chloride 26% in aqueous solution
Catalog # VWRS6039.5000
Supplier:  VWR Collection
CAS Number:  
Sodium chloride 26% in aqueous solution
Catalog # VWRS6039.5000
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    5 L
  • Seq:
    5190

Specifications

About this item

 :
 :58.44 g/mol
Density:1…1.2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-598-3
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477