Order Entry
Austria
ContactUsLinkComponent
Organic reference standard, Styrene 5,000 µg/ml in methanol, Dr Ehrenstorfer
Organic reference standard, Styrene 5,000 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERYA16982000ME
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, Styrene 5,000 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERYA16982000ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    100-42-5
  • Description:
    Styrene 5,000 µg/ml in methanol
  • Formula:
    C₈H₈
  • Description:
    Styrene

Specifications

About this item