Alginic acid, M.W. ~240,000 (from Macrocystis pyrifera (kelp)), off-white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:9005-32-7
:
Formula:(C₆H₈O₆)n
MDL Number:MFCD00081309
:176.13 g/mol
Melting Pt:300 °C
Storage Temperature:Ambient
EINECS:232-680-1
Frequently Bought Together