Order Entry
Austria
ContactUsLinkComponent
Alginic acid
Alginic acid
Catalog # MOLEM27060131
Supplier:  Molekula
CAS Number:  
Alginic acid
Catalog # MOLEM27060131
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :176.13 g/mol
Storage Temperature:Ambient
EINECS:232-680-1
CAS Number:9005-32-7
Formula:(C₆H₈O₆)n
MDL Number:MFCD00081309