Order Entry
Austria
ContactUsLinkComponent
Ethidium bromide 10 mg/ml in aqueous solution
Ethidium bromide 10 mg/ml in aqueous solution
Catalog # 40042.
Supplier:  Biotium
CAS Number:  
Ethidium bromide 10 mg/ml in aqueous solution
Catalog # 40042.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • Seq:
    2275

Specifications

About this item

 :
 :394.31 g/mol
EINECS:214-984-6
 :GHS06
 :GHS08
CAS Number:1239-45-8
Formula:C₂₁H₂₀BrN₃
MDL Number:MFCD00011724