Bromophenol blue 0.1% in water, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:115-39-9
:
Formula:C₁₉H₁₀Br₄O₅S
MDL Number:MFCD00005875
:669.97 g/mol
Density:1 g/cm³
Storage Temperature:Ambient
EINECS:204-086-2
Specification Test Results
- Concentration:(by UV) About 0.1%
- Infrared spectrum:Conforms
- Melting point:212 to 217 °C
- HPLC:≥97.5 %
- Specific optical rotation:+150° to +156° (20 °C, 589 nm) (C=1, dioxane)
Specification Test Results
Frequently Bought Together