Order Entry
Austria
ContactUsLinkComponent
Bromocresol green sodium salt in ACS Reagent grade water ≥90% (by HPLC), Sigma-Aldrich®
Zertifikate
undefined
Bromocresol green sodium salt in ACS Reagent grade water ≥90% (by HPLC), Sigma-Aldrich®

Spezifikation Testergebnisse

  • Appearance (Form):
    Powder or Crystals
  • Infrared spectrum:
    Conforms to Structure
  • Visual Interval 1, pH 3.8 = Yellow:
    Pass
  • Visual Interval 2, pH 4.5 = Green:
    Pass
  • Visual Interval 3, pH 5.4 = Blue:
    Pass
  • Clarity of solution not more than a faint trace of turbidity or insoluble matter c = 0.1 g/100 ml; Water:
    Conforms
  • Purity (HPLC):
    ≥90 %

Spezifikation Testergebnisse

Frequently Bought Together