Order Entry
Austria
ContactUsLinkComponent
 
Bromocresol green sodium salt, Millipore®
  1.01541.0010
 :  Merck
 :  
Bromocresol green sodium salt, Millipore®
  1.01541.0010
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

 

 

  • Molar Mass:
    719.99 g/mol

 

 

 :
 :225 °C
 :M-Clarity™ quality level = MQ200
 :720 g/mol
 :Ambient
 :263-657-4
 :62625-32-5
 :67763-24-0
 :C₂₁H₁₃Br₄NaO₅S
 :MFCD00148898