Order Entry
Austria
ContactUsLinkComponent
Bromocresol green sodium salt ACS
Bromocresol green sodium salt ACS
Catalog # 0548-50G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Bromocresol green sodium salt ACS
Catalog # 0548-50G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Insolubles Test:
    none detected
  • Visual Transition Interval (pH 3.8 – 5.4):
    Pass

Specification Test Results

About this item

 :
Melting Pt:225 °C
 :720 g/mol
Storage Temperature:Ambient
EINECS:263-657-4
CAS Number:62625-32-5
 :67763-24-0
Formula:C₂₁H₁₃Br₄NaO₅S
MDL Number:MFCD00148898