2,4-Dichloro-N-methylaniline
Marketsource
Supplier: Thermo Scientific
Jump to:
About this item
- CAS Number:35113-88-3
- Formula:C₇H₇Cl₂N
- MDL Number:MFCD00052853
- Molecular Weight:176.04 g/mol
- UN:2811
- ADR:6.1,III
2 Options Available Below
Product Details & Documents
- CAS Number:35113-88-3
- Formula:C₇H₇Cl₂N
- MDL Number:MFCD00052853
- Molecular Weight:176.04 g/mol
- UN:2811
- ADR:6.1,III
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource



