Herbicide reference standard, Chlorotoluron (3-(3-Chloro-p-tolyl)-1,1-dimethylurea) 500 µg/ml in methanol
Catalog # PROOSL20233
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:15545-48-9
- Description:Chlorotoluron (3-(3-Chloro-p-tolyl)-1,1-dimethylurea) 500 µg/ml in methanol
- Formula:C₁₀H₁₃ClN₂O
- Description:Chlorotoluron (3-(3-Chloro-p-tolyl)-1,1-dimethylurea)