ContactUsLinkComponent
Dynalon Two-Way Stopcocks with Hose Barb Connections
Dynalon Two-Way Stopcocks with Hose Barb Connections
Certificates
undefined
Dynalon Two-Way Stopcocks with Hose Barb Connections
Specifications
  • Connection type:
  • Barbed
  • Barbed
  • Barbed
  • Inlet port:
  • 16 mm Barb
  • 14 mm Barb
  • 10 mm Barb
  • Outlet port:
  • 16 mm Barb
  • 14 mm Barb
  • 10 mm Barb
  • Orifice:
  • 12,7 mm (0,5")
  • 10,7 mm (0,421")
  • 6,8 mm (0,268")
  • Tubing:
  • 16 mm
  • 14 mm
  • 10 mm
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • Wetted materials:
  • HDPE
  • HDPE
  • HDPE