ContactUsLinkComponent
Thermo Scientific™ Nalgene™ Two- and Three-Way Stopcocks with Hose Barb Connections
Thermo Scientific™ Nalgene™ Two- and Three-Way Stopcocks with Hose Barb Connections
Certificates
undefined
Thermo Scientific™ Nalgene™ Two- and Three-Way Stopcocks with Hose Barb Connections
Specifications
  • Connection type:
  • Barbed
  • Barbed
  • Barbed
  • Barbed
  • Inlet port:
  • 1/2" to 5/16" Barb
  • 1/2" to 5/16" Barb
  • 1/2" to 5/16" Barb
  • 1/4" to 5/16" Barb
  • Outlet port:
  • 1/2" to 5/16" Barb
  • 1/2" to 5/16" Barb
  • 1/2" to 5/16" Barb
  • 1/4" to 5/16"
  • Int. Ø tubing:
  • 0,25 - 0,3125"
  • 0,25 - 0,3125"
  • 0,25 - 0,3125"
  • 0,25 - 0,3125"
  • Orifice:
  • 2 mm (0,0787")
  • 4 mm (0,157")
  • 2 mm (0,0787")
  • 4 mm (0,157")
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • Wetted materials:
  • Polypropylene
  • Polypropylene
  • Polypropylene
  • Polypropylene