AP-18 ≥98% (by TLC)
Catalog # ENZOBMLCA2390025
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
Specification Test Results
- Purity:≥98% (TLC)
Specification Test Results
About this item
:
:209.7 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:55224-94-7
Formula:C₁₁H₁₂CINO