Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Bufalin
Bufalin ≥98% (by TLC)
Bufalin ≥98% (by TLC)
Bufalin ≥98% (by TLC)
Catalog # ENZOBMLAP3030010
Supplier:
Enzo Life Sciences
CAS Number:
Expand image
Bufalin ≥98% (by TLC)
Catalog # ENZOBMLAP3030010
Supplier:
Enzo Life Sciences
CAS Number:
Specifications
Pk:
10 mg
Seq:
2275
Specifications
About this item
Product Details
:
:
386.532 g/mol
CAS Number:
465-21-4
Formula:
C₂₄H₃₄O₄
MDL Number:
MFCD00056525
Product Details