Order Entry
Austria
ContactUsLinkComponent
 
Aflatoxin B2 ≥98% (by HPLC, TLC)
New Product
 
Aflatoxin B2 ≥98% (by HPLC, TLC)
New Product

 

  • Environmentally Preferable:

 

 

  • Purity:
    ≥98% (HPLC, TLC)
  • Source:
    Isolated from Aspergillus flavus

 

Frequently Bought Together