5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)] ≥95% fluorescent dye, for microbiology
Catalog # ENZOALX610030M025
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:557.467 g/mol
CAS Number:150347-59-4
:MFCD31812843
Formula:C₂₉H₁₉NO₁₁
MDL Number:MFCD00037466