Order Entry
Austria
ContactUsLinkComponent
D-(+)-Neopterin ≥98% (by HPLC)
D-(+)-Neopterin ≥98% (by HPLC)
Catalog # ENZOALX550126M050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
D-(+)-Neopterin ≥98% (by HPLC)
Catalog # ENZOALX550126M050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pack type:
    Vial, amber
  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Purity (HPLC):
    ≥98%
  • Appearance:
    Off-white to yellow solid

Specification Test Results

About this item

 :
 :253.218 g/mol
Merck Index:14,06462
EINECS:217-924-7
 :GHS07
Storage Temperature:Freezer
CAS Number:2009-64-5
Formula:C₉H₁₁N₅O₄
MDL Number:MFCD00042801