Order Entry
Austria
ContactUsLinkComponent
 
(+)-Himbacine ≥98%, powder
Catalog # ENZOALX550061M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
(+)-Himbacine ≥98%, powder
Catalog # ENZOALX550061M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

Specification Test Results

  • Purity:
    ≥98%
  • Source:
    Isolated from the Australian pine galbulimima baccata
  • Appearance:
    White to off-white powder

Specification Test Results

About this item

 :
 :345.5 g/mol
Storage Temperature:Ambient
CAS Number:6879-74-9
Formula:C₂₂H₃₅NO₂