Adenosine-3’,5’-cyclic Monophosphothioate, Rp-Isomer sodium salt ≥99% (by HPLC)
Catalog # ENZOALX480085M001
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 mg
- Seq:2070
Specifications
About this item
:
:344.3 · 23.0 g/mol
Storage Temperature:Freezer
CAS Number:73208-40-9
Formula:C₁₀H₁₁N₅O₅PS·Na