Adenosine 5'-diphosphate disodium salt (ADP disodium salt)
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:16178-48-6
:
Formula:C₁₀H₁₃N₅Na₂O₁₀P₂
MDL Number:MFCD00066635
:471.167 g/mol
EINECS:240-314-7
Frequently Bought Together