Order Entry
Austria
ContactUsLinkComponent
Irinotecan hydrochloride trihydrate ≥98% (by HPLC)
Irinotecan hydrochloride trihydrate ≥98% (by HPLC)
Catalog # ENZOALX430139M025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Irinotecan hydrochloride trihydrate ≥98% (by HPLC)
Catalog # ENZOALX430139M025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :677.19 g/mol (623.15 anhy)
CAS Number:136572-09-3
 :MFCD01862255
Formula:C₃₃H₃₈N₄O₆·HCl·3H₂O
MDL Number:MFCD01765731