Order Entry
Austria
ContactUsLinkComponent
 
HA14-1 ≥95% (by 1H-NMR)
 
undefined
HA14-1 ≥95% (by 1H-NMR)

 

  • Purity (¹H-NMR):
    ≥95%
  • Identity:
    Identity determined by IR, ¹H-NMR
  • Appearance:
    Off-white to light yellow powder

 

Frequently Bought Together