Spermine NONOate ≥97% (by NMR)
Supplier: ENZO LIFE SCIENCES
New Product
Certificates
About this item
CAS Number:136587-13-8
:
Formula:C₁₀H₂₆N₆O₂
:262.4 g/mol
:GHS07
Storage Temperature:Ultra-Low Temperature Freezer (–80°C)
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Purity:≥97% (NMR)
Specification Test Results
Frequently Bought Together