Order Entry
Austria
ContactUsLinkComponent
WS-12 ≥98% (by HPLC)
WS-12 ≥98% (by HPLC)
Catalog # ENZOALX420041M025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
WS-12 ≥98% (by HPLC)
Catalog # ENZOALX420041M025
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

Specification Test Results

  • Purity:
    ≥98% (HPLC)
  • Identity:
    Determined by ¹H-NMR and MS

Specification Test Results

About this item

 :
 :289.4 g/mol
 :GHS07
 :GHS08
Storage Temperature:Freezer
CAS Number:68489-09-8
Formula:C₁₈H₂₇NO₂
MDL Number:MFCD11100191