Order Entry
Austria
ContactUsLinkComponent
GW1929 ≥97% (by HPLC)
GW1929 ≥97% (by HPLC)
Catalog # ENZOALX420029M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
GW1929 ≥97% (by HPLC)
Catalog # ENZOALX420029M005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

Specification Test Results

  • Purity:
    ≥97% (HPLC)

Specification Test Results

About this item

 :
 :495.579 g/mol
Storage Temperature:Refrigerator
CAS Number:196808-24-9
Formula:C₃₀H₂₉N₃O₄