Order Entry
Austria
ContactUsLinkComponent
Blasticidin S hydrochloride (from Streptomyces griseochromogenes) ≥98% (by HPLC)
Blasticidin S hydrochloride (from Streptomyces griseochromogenes) ≥98% (by HPLC)
Catalog # ENZOALX380089M100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Blasticidin S hydrochloride (from Streptomyces griseochromogenes) ≥98% (by HPLC)
Catalog # ENZOALX380089M100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :458.904 g/mol
Storage Temperature:Refrigerator
CAS Number:3513-03-9
Formula:C₁₇H₂₇ClN₈O₅
MDL Number:MFCD02091640