Rifampicin
Glass bottle for solids, 100 mg
Catalog # ENZOALX380071M100
Supplier: Enzo Life Sciences
Jump to:
Product Details & Documents
Synonyms: Rifampin, 3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifamycin AMP
- CAS Number:13292-46-1
- Formula:C₄₃H₅₈N₄O₁₂
- MDL Number:MFCD00151389
- Molecular Weight:822.95 g/mol
- Merck Index:12,08382
- Melting Pt:183…188 °C
- EINECS:236-312-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



