Order Entry
Austria
ContactUsLinkComponent
 
Puerarin ≥98% (by HPLC)
Catalog # ENZOALX350249M005
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  
Puerarin ≥98% (by HPLC)
Catalog # ENZOALX350249M005
Supplier:  ENZO LIFE SCIENCES
New Product
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 8-(beta-D-Glucopyranosyl)-4',7-dihydroxyisoflavone
 :
 :416.38 g/mol
CAS Number:3681-99-0
Formula:C₂₁H₂₀O₉
MDL Number:MFCD00076007