Order Entry
Austria
ContactUsLinkComponent
Sibutramine hydrochloride monohydrate ≥98% (by HPLC)
Sibutramine hydrochloride monohydrate ≥98% (by HPLC)
Catalog # ENZOALX350111G001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Sibutramine hydrochloride monohydrate ≥98% (by HPLC)
Catalog # ENZOALX350111G001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Purity:
    ≥98% (HPLC)

Specification Test Results

About this item

 :
 :334.4 g/mol
Merck Index:14,08485
Melting Pt:190…201 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:125494-59-9
Formula:C₁₇H₂₆ClN·HCl·H₂O