Order Entry
Austria
ContactUsLinkComponent
Deoxymannojirimycin hydrochloride ≥98% (by TLC)
Deoxymannojirimycin hydrochloride ≥98% (by TLC)
Catalog # ENZOBMLS1090005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Deoxymannojirimycin hydrochloride ≥98% (by TLC)
Catalog # ENZOBMLS1090005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :199.6 g/mol
Storage Temperature:Freezer
CAS Number:84444-90-6
Formula:C₆H₁₃NO₄·HCl