Order Entry
Austria
ContactUsLinkComponent
Deoxyfuconojirimycin hydrochloride ≥98% (by TLC)
Deoxyfuconojirimycin hydrochloride ≥98% (by TLC)
Catalog # ENZOBMLS1080005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Deoxyfuconojirimycin hydrochloride ≥98% (by TLC)
Catalog # ENZOBMLS1080005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :183.6 g/mol
Storage Temperature:Refrigerator
CAS Number:210174-73-5
Formula:C₆H₁₃NO₃·HCl