ITSA-1 ≥98% (by TLC)
Catalog # ENZOBMLGR3500025
Supplier: ENZO LIFE SCIENCES
New Product
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
- Environmentally Preferable:
Specifications
About this item
Synonyms: (1H-Benzo[d][1,2,3]triazol-1-yl)(2,4-dichlorophenyl)methanone
:
:292.1202 g/mol
CAS Number:200626-61-5
Formula:C₁₃H₇Cl₂N₃O
MDL Number:MFCD00195103