Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Eicosapentaenoyl ethanolamide
Eicosapentaenoyl ethanolamide ≥98% (by TLC)
Eicosapentaenoyl ethanolamide ≥98% (by TLC)
ENZOBMLFA0470025
:
ENZO LIFE SCIENCES
New Product
:
Eicosapentaenoyl ethanolamide ≥98% (by TLC)
ENZOBMLFA0470025
:
ENZO LIFE SCIENCES
New Product
:
Pk:
25 mg
Seq:
2315
Environmentally Preferable:
:
:
345.5 g/mol
:
109001-03-8